Få mere at vide
Flunarizine EP Impurity C (as Dihydrochloride), Mikromol™
Beskrivelse
Flunarizine EP Impurity C (as Dihydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-[(RS)-(2-Fluorophenyl)(4-fluorophenyl)methyl]-4-[(2E)-3-phenylprop-2-enyl]piperazine Dihydrochloride |
| CAS | 2734416-95-4 |
| Analyt- eller komponentnavne | 1-[(RS)-(2-Fluorophenyl)(4-fluorophenyl)methyl]-4-[(2E)-3-phenylprop-2-enyl]piperazine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C26 H26 F2 N2 . 2[HCl] |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic Acid, Flunarizine Dihydrochloride Imp. C (EP), Flunarizine Imp. C (EP), 1-[(RS)-(2-Fluorophenyl)(4-fluorophenyl)methyl]-4-[(2E)-3-phenylprop-2-enyl]piperazine Dihydrochloride, Flunarizine Dihydrochloride Imp. C (EP) as Dihydrochloride, 4-Defluoro-2-fluoro-flunarizine dihydrochloride, Cloxacillin Related Compound D, Cloxacillin Sodium Impurity D, Cloxacillin Impurity D, Flunarizine Impurity C Dihydrochloride, (E)-1-[(4-Fluoroohenyl)(2-fluorophenyl)methyl]-4-(cinnamyl)piperazine Dihydrochloride, 1-cinnamyl-4-((2-fluorophenyl)(4-fluorophenyl)methyl)piperazine dihydrochloride |
| InChI formel | InChI=1S/C26H26F2N2.2ClH/c27-23-14-12-22(13-15-23)26(24-10-4-5-11-25(24)28)30-19-17-29(18-20-30)16-6-9-21-7-2-1-3-8-21;;/h1-15,26H,16-20H2;2*1H/b9-6+;; |
| SMIL | FC1=CC=C(C=C1)C(C2=CC=CC=C2F)N3CCN(C/C=C/C4=CC=CC=C4)CC3.Cl.Cl |
| IUPAC navn | 1-[(2-fluorophenyl)-(4-fluorophenyl)methyl]-4-[(E)-3-phenylprop-2-enyl]piperazine;dihydrochloride |
| Vis mere |