Få mere at vide
Flumequine EP Impurity B, Mikromol™
Artikelnummer. 30051601
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051601 100 mg
30050408 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051601 Leverandør Mikromol Leverandørnr. MM0842.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Flumequine EP Impurity B
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Flumequine Ethyl Ester
CAS 42835-47-2
Analyt- eller komponentnavne Flumequine Ethyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C16 H16 F N O3
Certificeringer/compliance ISO 9001
Synonym Ethyl (RS)-9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-benzo[i,j]quinolizine-2-carboxylate (Flumequine Ethyl Ester), Flumequine Imp. B (EP), 9-fluoro-6,7-dihydro-5-methyl-1-oxo-1H,5H-Benzo[ij]quinolizine-2-carboxylic acid ethyl ester, Flumequine ethyl ester, Flumequine Impurity B
InChI formel InChI=1S/C16H16FNO3/c1-3-21-16(20)13-8-18-9(2)4-5-10-6-11(17)7-12(14(10)18)15(13)19/h6-9H,3-5H2,1-2H3
SMIL CCOC(=O)C1=CN2C(C)CCc3cc(F)cc(C1=O)c23
IUPAC navn Ethyl (RS)-9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-benzo[i,j]quinolizine-2-carboxylate
Molekylvægt (g/mol) 289.3
Mængde 100 mg
Formel vægt 289.1114 g/mol