Få mere at vide
Flumazenil EP Impurity D, Mikromol™
Artikelnummer. 30052605
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052605 100 mg
30050819 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052605 Leverandør Mikromol Leverandørnr. MM0762.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Flumazenil EP Impurity D
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Fluoro-4-methyl-3,4-dihydro-1H-1,4-benzodiazepine-2,5-dione
CAS 78755-80-3
Analyt- eller komponentnavne 7-Fluoro-4-methyl-3,4-dihydro-1H-1,4-benzodiazepine-2,5-dione
Forsendelsestilstand Room Temperature
Molekylær formel C10 H9 F N2 O2
Certificeringer/compliance ISO 9001
Synonym 7-Fluoro-3,4-dihydro-4-methyl-1H-1,4-benzodiazepine-2,5-dione
InChI formel InChI=1S/C10H9FN2O2/c1-13-5-9(14)12-8-3-2-6(11)4-7(8)10(13)15/h2-4H,5H2,1H3,(H,12,14)
SMIL CN1CC(=O)Nc2ccc(F)cc2C1=O
IUPAC navn 7-fluoro-4-methyl-1,3-dihydro-1,4-benzodiazepine-2,5-dione
Molekylvægt (g/mol) 208.19
Mængde 100 mg
Formel vægt 208.192 g/mol