Få mere at vide
Thermo Scientific™ FluidEase™ Pro ClipTip™ Electronic Multichannel Pipettes
Artikelnummer. p-200174111 Shop alle Thermo Scientific produkter
Skift visning
Volumen (metrisk):
0.5 to 10 μL
1 to 30 μL
10 to 300 μL
2.5 to 100 μL
25 to 1250 μL
3 to 125 μL
5 to 200 μL
Antal kanaler:
8
12
16
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Volumen (metrisk) Antal kanaler
31891611 0.5 to 10 μL 8
31891594 2.5 to 100 μL 8
31891602 5 to 200 μL 8
31891597 10 to 300 μL 8
31891600 25 to 1250 μL 8
31891612 0.5 to 10 μL 12
31891606 2.5 to 100 μL 12
31891609 5 to 200 μL 12
31891593 10 to 300 μL 12
31891601 25 to 1250 μL 12
31891604 0.5 to 10 μL 16
31891595 1 to 30 μL 16
31891596 3 to 125 μL 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Artikelnummer. 31891611 Leverandør Thermo Scientific™ Leverandørnr. 4676010
Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic multichannel pipettes enable effortless 96- and 384-well plate filling with touchscreen control, ultra-wide speed selection, and secure ClipTip™ sealing for reproducible, high-throughput workflows in shared labs.
Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic multichannel pipettes are designed to simplify and standardize plate-based workflows. A large touchscreen interface and ultra-wide speed selection support precise aspiration and dispense across liquid types. Secure ClipTip™ sealing, effortless column-to-row transitions, preset and advanced programming modes, and password-protected settings enable reproducible, high-throughput plate filling across shared laboratory environments
Features:
  • Large Touchscreen User Interface: A high-resolution touchscreen provides direct, visual interaction for setup and execution, reducing training time and minimizing setup errors compared to button- or joystick-based navigation.
  • Ultra-Wide Speed Selection: One of the broadest aspiration and dispense speed ranges available supports gentle handling of sensitive samples, precise control of viscous liquids, and faster execution for time-critical workflows.
  • Simple Presets and Advanced Programming: Easy-to-use preset modes support fast onboarding for new or occasional users, while advanced step programming enables complex, multi-step protocols for demanding applications.
  • Index-Finger Operation with Increased Rotational Flexibility: Retains Thermo Fisher–recognized index-finger operation with a wider angle of rotation at the finger rest to accommodate different hand sizes, grip angles, and working positions.
  • Secure ClipTip™ Technology: ClipTip™ tips lock and seal in place until intentional ejection, helping ensure consistent volume delivery and reduced variability from dispense to dispense.
  • Electronic Tip Ejection with Safe-to-Eject Illumination: Electronic tip ejection reduces physical effort and includes LED illumination to indicate when ejection is possible, providing ergonomic benefit and added user security.
  • Password Protection and User Profiles: Password-protect critical settings and protocols and support up to five user profiles to enable standardized operation in shared and regulated laboratory environments.
  • Custom Calibration Options: Enable calibration to customer-specific liquids or environmental conditions to align performance with real laboratory workflows.
  • Service and Maintenance Tracking: Built-in service reminders, calibration logs, and usage tracking support instrument management and compliance documentation over time.
TRUSTED_SUSTAINABILITY
Tekniske data
Justerbar spidsafstand No
Autoklaverbar Not Autoclavable
Farve Pink
Engangs Reusable
Til brug med ClipTip 12.5
Model- Bluetooth
Antal kanaler 8
Programmerbarhed Yes
Sterilitet Non-sterile
Type Electronic Pipette
Volumen (metrisk) 0.5 to 10 μL
Vise Touchscreen
Finger stil Ergonomic Index Finger
Flersproget brugergrænseflade Yes
garanti 1 Year, 2 Years with Registration