Få mere at vide
Thermo Scientific™ Fluid Transfer Assembly
Specialty Fluid Transfer Assembly products integrate easily within your existing process and equipment to maximize productivity, reduce cross-contamination and keep your process running smoothly.
Brand:  Thermo Scientific™ SS00062-I
 Se flere versioner af dette produkt
Produktkode 15969916
  • Ring for pris / 25 enheder
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Specialized connectors and tubing in our ready-designed assemblies provide an off-the-shelf solution that addresses many fluid transfer needs while delivering efficiency and increasing process integrity.
  • End treatments facilitate simple interconnection with existing process and equipment
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
91.4cm of 12.7cm ID Silicone Tubing with 3.8cm Sanitary and 1.3cm MPX Body Connections