missing translation for 'onlineSavingsMsg'
Få mere at vide

Flufenpyr-ethyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Artikelnummer. 30313705
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313705 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30313705 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA13713000AL100

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Flufenpyr-ethyl
CAS 188489-07-8
Analyt- eller komponentnavne Flufenpyr-ethyl
Forsendelsestilstand Room Temperature
Molekylær formel C16 H13 Cl F4 N2 O4
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17034
InChI formel InChI=1S/C16H13ClF4N2O4/c1-3-26-14(24)7-27-13-5-12(11(18)4-10(13)17)23-15(25)8(2)9(6-22-23)16(19,20)21/h4-6H,3,7H2,1-2H3
SMIL CCOC(=O)COc1cc(N2N=CC(=C(C)C2=O)C(F)(F)F)c(F)cc1Cl
IUPAC navn ethyl 2-[2-chloro-4-fluoro-5-[5-methyl-6-oxo-4-(trifluoromethyl)pyridazin-1-yl]phenoxy]acetate
Opløsningsmiddel eller Matrix Acetonitrile
Molekylvægt (g/mol) 408.73
Mængde 1 ml
Formel vægt 408.05 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.