missing translation for 'onlineSavingsMsg'
Få mere at vide

Flucarbazone-N-desmethyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Artikelnummer. 30309873
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309873 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30309873 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA13685200AL100

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Flucarbazone-N-desmethyl
CAS 749198-26-3
Analyt- eller komponentnavne Flucarbazone-N-desmethyl
Forsendelsestilstand +20°C
Molekylær formel C11 H9 F3 N4 O6 S
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17034
Synonym 1H-1,2,4-Triazole-1-carboxamide, 2,5-dihydro-3-methoxy-5-oxo-N-[[2-(trifluoromethoxy)phenyl]sulfonyl]-
InChI formel InChI=1S/C11H9F3N4O6S/c1-23-8-15-9(19)18(16-8)10(20)17-25(21,22)7-5-3-2-4-6(7)24-11(12,13)14/h2-5H,1H3,(H,17,20)(H,15,16,19)
SMIL COC1=NN(C(=O)NS(=O)(=O)c2ccccc2OC(F)(F)F)C(=O)N1
IUPAC navn 3-methoxy-5-oxo-N-[2-(trifluoromethoxy)phenyl]sulfonyl-4H-1,2,4-triazole-1-carboxamide
Opløsningsmiddel eller Matrix Acetonitrile
Molekylvægt (g/mol) 382.27
Mængde 1 ml
Formel vægt 382.0195 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.