Learn More
Cole-Parmer Flow Through Tubing Weight, PTFE
Product Code. 30053089 Shop All Cole-Parmer Products
Change view
Click to view available options
Tubing Size:
3/16 in. to 1/4 in.
5/16 in. to 7/16 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Unit Size
30053089 3/16 in. to 1/4 in. Each
30054041 5/16 in. to 7/16 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30053089 Supplier Cole-Parmer Supplier No. 5011798
  • ONLINE EXCLUSIVE PRICE
    672.00 DKK
    655.80 DKK / Pakke med 1
    Spare 16.20 DKK
    2% Slukket
View all promotions
Forventet forsendelse: 21-12-2026
Prevent tubing from coiling or floating during fluid transfer applications
Tapered hose barb fitting easily inserts into the inner diameter (ID) of flexible tubing. Keep tubing submerged in liquid, preventing floatation, and ensuring uninterrupted fluid flow. Constructed of chemically inert PTFE. Autoclavable—allowing for easy sterilization and reuse
Specifications
Tubing Size 3/16 in. to 1/4 in.