Få mere at vide
Florfenicol, 98%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J66995.03
| Mængde | 1g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
Kemiske identifikatorer
| 73231-34-2 | |
| 358.205 | |
| AYIRNRDRBQJXIF-NXEZZACHSA-N | |
| CHEBI:87185 | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O |
| C12H14Cl2FNO4S | |
| MFCD00864834 | |
| 114811 | |
| 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
Tekniske data
| Florfenicol | |
| White | |
| >110°C (230°F) | |
| MFCD00864834 | |
| 14,4109 | |
| AYIRNRDRBQJXIF-NXEZZACHSA-N | |
| 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide | |
| 114811 | |
| 358.21 |
| 73231-34-2 | |
| 1g | |
| C12H14Cl2FNO4S | |
| 618°C | |
| Soluble in ethanol to 25mM and in DMSO to 100mM | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O | |
| 358.205 | |
| CHEBI:87185 | |
| 98% |