Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UN160
Flexible temperature sensor for positioning in chamber or in load.
Brand:  Memmert™ UN160H8
Product Code. 17579885
  • 1690.00 DKK / Each
Only null left
Estimated Shipment: 24-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens
Specifications
Specifications
UN160 Oven
Temperature Sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Flexible Pt100 Temperature Sensor for UN160 > For Use With: UN160