Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF55
Flexible temperature sensor for positioning in chamber or in load.
Brand:  Memmert™ UF55H8
1641.41 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17549465
  • 1690.00 DKK / Each
Only null left
Estimated Shipment: 03-09-2025
Description
  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens
Specifications
Specifications
UF55 Oven
Temperature Sensor
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Flexible Pt100 Temperature Sensor for UF55 > For Use With: UF55