Learn More
Flavoxate hydrochloride, MedChemExpress
Product Code. 16452229
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452229 1 g 1g
16448366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16452229 Supplier Medchem Express Supplier No. HYB0549A
 Restricted Item
Forventet forsendelse: 30-11-2026
MedChemExpress Flavoxate hydrochloride is a potent and competitive phosphodiesterase (PDE) inhibitor. Flavoxate hydrochloride is an antispasmodic agent and muscarinic mAChR antagonist. Flavoxate hydrochloride shows moderate calcium antagonistic activity and local anesthetic effect. Flavoxate hydrochloride can be used for the research of overactive bladder (OAB) and lower urinary tract infections.
Flavoxate hydrochloride is a potent and competitive phosphodiesterase (PDE) inhibitor. Flavoxate hydrochloride is an antispasmodic agent and muscarinic mAChR antagonist. Flavoxate hydrochloride shows moderate calcium antagonistic activity and local anesthetic effect. Flavoxate hydrochloride can be used for the research of overactive bladder (OAB) and lower urinary tract infections.
Specifications
Chemical Name or Material Flavoxate hydrochloride
CAS 3717-88-2
Color White
Purity Grade Notes Research
Molecular Formula C24H26ClNO4
Quantity 1 g
Synonym Rec-7-0040 DW61
Solubility Information H2O : 5 mg/mL (11.68 mM; Need ultrasonic) ∣DMSO : 3.33 mg/mL (7.78 mM; Need ultrasonic)
SMILES O=C(C1=C2C(C(C(C)=C(C3=CC=CC=C3)O2)=O)=CC=C1)OCCN4CCCCC4.Cl
Molecular Weight (g/mol) 427.92
Formula Weight 427.92
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation