missing translation for 'onlineSavingsMsg'
Få mere at vide

Flavoxathydrochlorid, LoGiCal

Artikelnummer. 30461284
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30461284 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30461284 Leverandør LoGiCal Leverandørnr. LGCFOR0156.00
 Begrænset vare
Forventet forsendelse: 30-11-2026
Læg i indkøbskurv
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCals certificerede referencematerialer som smarte, native løsninger og stabile isotop-mærkede løsninger, med ekspertdokumentation og support. Fra retsmedicinske til kliniske laboratorier – vælg LoGiCal for selvtillid, overholdelse og analytisk ekspertise.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Flavoxate Hydrochloride
CAS 3717-88-2
Analyt- eller komponentnavne Flavoxate Hydrochloride
Forsendelsestilstand Stuetemperatur
Molekylær formel C24 H25 N O4 . Cl H
Certificeringer/compliance ISO 9001
Synonym Flavoxate Hydrochloride, 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid 2-(1-piperidinyl)ethyl ester hydrochloride, 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid 2-piperidinoethyl ester hydrochloride, 1-Piperidineethanol 3-methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate (ester) hydrochloride, Bladderon, DW 61, Flavoxate hydrochloride, Genurin, NSC 114649, Patricin, Peflate, Piperidinoethyl-3-methylflavone-8-carboxylate hydrochloride, Rec 7/0040, Spasmal, Spasuret, Urispas
InChI formel InChI=1S/C24H25NO4.ClH/c1-17-21(26)19-11-8-12-20(23(19)29-22(17)18-9-4-2-5-10-18)24(27)28-16-15-25-13-6-3-7-14-25;/h2,4-5,8-12H,3,6-7,13-16H2,1H3;1H
Løsningstype Fedt
SMIL Cl.CC1=C(Oc2c(cccc2C1=O)C(=O)OCCN3CCCCC3)c4ccccc4
IUPAC navn 2-piperidin-1-ylethyl 3-methyl-4-oxo-2-fenylchromen-8-carboxylat; hydrochlorid
Industry Type Klinisk testning
Molekylvægt (g/mol) 427.92
Mængde 10 mg
Formel vægt 427,155 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.