Learn More
Lenz Laborglasintrumente Flat Bottom Flasks, Wide Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07050074
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11963831
  • 4690.00 DKK / Pack of 10
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN ISO 24450
  • With beaded rim
  • Material: DURAN™
Specifications
Specifications
2 L
Duran Glass
240 mm