Få mere at vide
Eppendorf Flask Clamp for Incubator Shaker CS220, HTD Compatible
Artikelnummer. 30280819 Shop alle Eppendorf produkter
Holder:
1 l kolber
100/125 ml kolber
2 l kolber
2,8 l kolber
200/250 ml kolber
3/5 l kolber
500 ml kolber
Artikelnummer. 30280819
Brand: Eppendorf 6740070225
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Eppendorf flask clamps secure a wide range of vessels to the shaker platform and are constructed of a single piece stainless-steel for added strength and safety. Flask clamps come complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
  • For Erlenmeyer flasks (standard, narrow/wide neck), Corning™, Thomson™, or Nalgene™
  • Fernbach
  • 180°C resistant
  • Autoclavable
  • Stainless steel
  • Includes M4x7 screws
TRUSTED_SUSTAINABILITY
Tekniske data
Holder 3/5 l kolber
Produkttype Lab Shaker Clamp
Materiale Stainless Steel
Til brug med (udstyr) CellXpert CS220 CO2 Incubator Shaker
Inkluderer M4x7 screws
Mængde 1 Ea.