Learn More
Flash Point Standard, COC ASTM D92 ( 153.0 °C nominal value), Paragon Scientific™
Product Code. 16000040
3 x 80 mL, Pack of 3
Click to view available options
Quantity:
3 x 80 mL
Unit Size:
Pack of 3
These are certified reference materials characterized by a method specific multi-laboratory study in accordance with ASTM D92. These materials are suitable for verifying the performance of an instrument and/or procedure as specified in this standard.
  • Certified Reference Material for annual verification checks in accordance with ASTM D92.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Packaged in 3 x 80 mL bottles – ‘One shot’, one bottle per test ensures zero contamination/ high-end volatile loss.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
Quantity 3 x 80 mL
Nominal values quoted. Certified Values will vary from batch to batch.
Product Content Correction
Product Title
Flash Point Standard, COC ASTM D92 ( 153.0 °C nominal value), Paragon Scientific™ >