Learn More
Trajan Scientific and Medical™ Gas Tight Fixed Needle Autosampler Syringe, Side Hole Tip
Designed to provide improved solvent resistance, greater temperature range and increased operational smoothness.
Brand:  Trajan Scientific and Medical™ 007890
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13166105
  • 1376.00 DKK / Each
Estimated Shipment
14-06-2021
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
CTC RTC and Thermo RSH Syringe
0.32 mm
PTFE
Fixed Needle
Side Hole
23 G
0.63 mm
500 µL
57 mm