Få mere at vide
3M™ Fixed Length Breathing Tube SS-BT-23
Artikelnummer. 17736973 Shop alle 3M produkter
:
Breathing Tube; For Use With: 3M™ PAPR Systems
Artikelnummer. 17736973
Brand: 3M™ 7100236823
3M™ Fixed Length Breathing Tube SS-BT-23 is a fixed-length polyurethane breathing tube for use with approved 3M™ Powered Air Systems.
  • Fixed-length breathing tube made from polyurethane
  • Approved for use with 3M™ Powered Air Respirator PF-600E
  • Compatible with specific 3M™ Versaflo™ headtops when using the 3M™ Adapter Assembly QRS Thread ADP-03.
  • Protective breathing tube covers available to lengthen service life of breathing tubes