Learn More
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid
For use with Centrifuge 5430 and Centrifuge 5430 R
Brand: Eppendorf™ 5427752005
6880.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Aerosol-tight Eppendorf QuickLock rotors close with only 1/4 of a turn
- Saves time during every day use
- Takes repetitive stress off your wrist
Specifications
Model: FA-45-24-11 | |
24 Spin Columns | |
13,200 rpm | |
Fixed | |
24 |
Fixed Angle Rotor | |
19,090 x g | |
5430/5430R Centrifuges | |
Aerosol rotor lid |
Product Content Correction
Product Title
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid > Model: FA-45-24-11
Spot an opportunity for improvement?Share a Content Correction