Learn More
FITC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA128347
Additional Details : Weight : 0.01000kg
Description
PA1-28347 detects FITC from tagged samples. PA1-28347 has been successfully used in ELISA and Western blot applications. The PA1-28347 immunogen is FITC. Store at 4oC short term. For extended storage aliquot and store at -20oC or below. Avoid Freeze/Thaw Cycles.
FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs μgtraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.Specifications
FITC | |
Polyclonal | |
Unconjugated | |
Fluorescein | |
FITC | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Chemical | |
Antibody | |
IgG |