Få mere at vide
Fisherbrand™ Labtool II™ and Mini-Labtool™
Shop alle Fisherbrand produkter
Længde (engelsk):
3.54 in.
Vægt (engelsk):
0.43 lb.
Pakningsstørrelse:
Hver
Beskrivelse
Stainless-steel tools with aluminum handles open bottles; cut stoppers, tubing and wires; and much more.
Labtool II
- Includes pliers/gripper, wire stripper, wire cutter, ruler, small/large knife, serrated knife, punch, flat head and Phillips screwdrivers, bottle opener, needle, saw and serrated stripper
- Closed length: 3.5 in. (9cm)
- Weight: 7 oz. (200g)
Mini-Labtool
- Includes pliers/gripper, wire stripper, wire cutter, knife, flat head and Phillips screwdrivers, scissors, file, saw, bottle and can openers and key chain
- Closed length: 2.5 in. (6.5cm)
- Weight: 2.75 oz. (79g)
Tekniske data
Tekniske data
| Produkttype | Labtool II |
| Længde (engelsk) | 3.54 in. |
| Længde (metrisk) | 9 cm |
| Materiale | Stainless Steel |
| Inkluderer | Pliers/gripper, wire stripper, wire cutter, ruler, small/large knife, serrated knife, punch, flat head and Phillips™ screwdrivers, bottle opener, needle, saw and serrated stripper |
| Vægt (engelsk) | 0.43 lb. |
| Vægt (metrisk) | 0.2 kg |