Få mere at vide
Fisherbrand™ Fisher Branded PurSwab 5 in. Rectangular Foam Swab w/Polypropylene Handle
Rectangular Foam Tipped Applicator with Plastic Handle
Brand: Fisherbrand™ 149603L
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Flexible mini-tip foam applicator with black plastic handle. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.
Used in controlled environments, cleanrooms and industrial settings. Coating rechargeable copier cartridges. Applying solutions or solvents. General cleaning swab. Tip is thermally bonded to handle so there is no contaminating adhesives used. Tip is lint and residue free. Can be used with IPA and DI water or dry. Made in USA