Learn More
Fisherbrand™ Disposable Polypropylene Bouffant Cap
Single-use cap provides basic protection
Brand:  Fisherbrand™
Additional Details : Weight : 2.50000kg
 View more versions of this product
Product Code. 15401942
  • 280.00 DKK / Pack of 1000
Estimated Shipment
31-01-2018
Add to basket
Description
Description
  • Disposable
  • 14gsm polypropylene
  • Dia.: 21 in. (53.3cm)
  • Latex-free
  • White
Specifications
Specifications
White
Yes
Polypropylene
21 in. (53.3cm)
Videos
Documentation