Learn More
Fisherbrand™ Protective Disposable Face Masks
Provides convenient protection
Brand:  Fisherbrand™
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15471932
  • 1950.00 DKK / Pack of 500
Estimated Shipment date
from Supplier 25-05-2020
Add to basket
Description
Description
  • Advanced protection
  • Disposable, single use
  • Choice of ear loops or tie closure
  • Nonwoven polypropylene outer layer
  • Thermally bonded nonwoven polypropylene inner layer
  • Latex-free
  • Choice of blue or white
  • Universal size/fit
Specifications
Specifications
White
Yes
Yes
50/pack 10 packs per case
3-ply
Nonwoven Polypropylene (outer and inner) / Meltblown Polypropylene (filter)
Earloops
PP (Polypropylene)
Universal