Learn More
Fisherbrand™ 3/8 in. Dot Labels
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
Brand: Fisherbrand™
Additional Details : Weight : 0.18140kg
Description
- Labels can be printed/written on and will not bleed, smudge or smear - perfect for identifying specimens after long periods of cryogenic storage
Specifications
Gold | |
Non-sterile | |
0.37 in. |
Dot label roll | |
1000/Roll | |
9.55 mm |