Learn More
Fisherbrand™ DNA Sequencing Systems: Combs for Model FB-SEQ-3545
DNA Sequencing Systems Combs for model FB-SEQ-3545
Brand:  Fisherbrand™
Additional Details : Weight : 0.00800kg
 View more versions of this product
Product Code. 11987505
  • 361.00 DKK / Each
Estimated Shipment: 29-08-2024
to see stock.
Description
Description
For use with model FB-SEQ-3545
Specifications
Specifications
Aluminum backed sequencer
50 cm
17.7 in.
45 cm
40
15μL
0.68 cm
19.6 in.
9 in.
23 cm
Comb
0.4 mm
13.7 in.
23 cm