Learn More
Fisherbrand™ Vortex Mixer Accessories Kits
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 15756939
  • 1420.00 DKK / Each
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
Foam Microtube and Microwell Plate Holder Kit with Retainer
  • Includes one microtube holder, one microwell plate holder, and one insert retainer (Cat. Nos. 02-215-386, -387, -391)
Foam Tube Holder Kit with Retainer
  • Iincludes 3 tube holders (for 9-13mm, 14-19mm, and 20-25mm tubes), one flat foam insert, two vessel harnesses and one insert retainer (Cat. Nos. 02-215-393, -394, -395, -397, -392, -391)
Microtube Kit:
  • Mixes (48) 0.25 to 2mL micro tubes; includes foam insert and insert retainer
Microwell Plate Kit:
  • For mixing 96-well plates or deep well blocks; includes foam insert and insert retainer
Specifications
Specifications
Foam Tube Holder Kit with Retainer
Vortex mixer
Holder for 9 to 13mm, 14 to 19mm and 20 to 25mm tubes, flat form insert, vessel harness, insert retainer