Learn More
Fisherbrand™ Standard Exhaust Oil Filters
As supplied with Fisher Scientific Maxima C Plus pumps
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10070011
  • 1252.27 DKK / Each
Estimated Shipment: 05-07-2022
to see stock.
Add to basket
Description
Description
As supplied with Fisher Scientific Maxima C Plus pumps
These compact, round filters simply screw into threaded exhaust port to collect oil expelled during operation. A replaceable filter element within the oil filter separates the oil into particles no larger than 0.3μm, and drops collected oil back into the oil case.
  • Filter element is easily replaced
  • Reorder number located on filter canister
1For M4C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
2For M4C or M8C vacuum pumps designed after August 1999 (S/N FKK090000 and newer).
Specifications
Specifications
2-1/2 in.
Model M8C vacuum pump
11.73 cm
63.5 mm
4.62 in.
Standard Exhaust Oil Filter