Learn More
Fisherbrand™ Standard Exhaust Oil Filters
As supplied with Fisher Scientific Maxima C Plus pumps
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
As supplied with Fisher Scientific Maxima C Plus pumps
These compact, round filters simply screw into threaded exhaust port to collect oil expelled during operation. A replaceable filter element within the oil filter separates the oil into particles no larger than 0.3μm, and drops collected oil back into the oil case.
- Filter element is easily replaced
- Reorder number located on filter canister
1For M4C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
2For M4C or M8C vacuum pumps designed after August 1999 (S/N FKK090000 and newer).
Specifications
2-1/2 in. | |
Model M2C and M4C vacuum pump | |
6.35 cm |
63.5 mm | |
2.5 in. | |
Standard Exhaust Oil Filter |