Description
- Start, stop and reset are all achieved with a single button
- Provides single-action timing up to 15 minutes with 0.1-second resolution
- Extra-tough metal housing
Specifications
Specifications
| Produkttype | Single-action Stopwatch |
| Tidsområde | 15 min. |
| Opløsning | 0.1 sec. |
| Diameter (metrisk) Indvendig | 5.08 x 1.27 cm |
| Vægt (engelsk) | 3 oz. |
| Vægt (metrisk) | 85.04 g |
| Til brug med (applikation) | Lab and plant use, provides single-action timing up to +15 min. with 0.1 sec. resolution |