Få mere at vide
Fisherbrand™ Luftdrevne analoge omrørere
Shop alle Fisherbrand produkter
Hastighed:
10.000 omdr./min.
1000 omdr./min.
1200 omdr./min.
3000 omdr./min.
Volumen:
10 l
20 l
50 l
Moment:
45
203
294
633
Pakningsstørrelse:
Hver
Beskrivelse
Indeholder
- A 9/16 in. dia. x 10 in. L support rod Four ft. air hose with double fittings for connection to air supply
- Needle valve allows variation in mixing speed
- Special muffler ensures quieter operation
- Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
- Can be used with flammable chemicals for explosion proof mixing
Ordreinfo
Aksel, propel, stativ og klemme fås separat
Tekniske data
Tekniske data
| Hastighed | 3000 omdr./min. |
| Volumen | 50 l |
| Vise | Analog |
| Spænding | 0/1 V |
| Diameter (engelsk) | 0.5 in. |
| Diameter (metrisk) aksel | 1.27 cm |
| Højde (engelsk) | 9 in. |
| Højde (metrisk) | 23 cm |
| Længde (engelsk) | 14 in. |
| Længde (metrisk) | 36 cm |
| Vis mere |