Få mere at vide
Fisherbrand™ Accessories for PowerGen™ High Throughput Homogenizers
Artikelnummer. 12397209 Shop alle Fisherbrand produkter
Diameter (engelsk):
3/20 tommer
Pakningsstørrelse:
Pakke med 5000
Artikelnummer. 12397209
For use with PowerGen™ High Throughput Homogenizers.
  • Vials are supplied with caps and are available with or without stainless-steel mixing beads
  • Magnetic pipet tips are used to remove beads from microplates
  • Bead dispenser places beads into plates easily and efficiently
Tekniske data
Produkttype Grinding Beads
Diameter (engelsk) 3/20 tommer
Materiale Stainless Steel
Til brug med (udstyr) PowerGen™ high throughput homogenizer