Få mere at vide
Fisetin-2',5',6,6',8-d5, CDN
Artikelnummer. 30324597
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324597 0.01 g
30318591 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324597 Leverandør CDN Leverandørnr. CDND80840.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fisetin-2',5',6,6',8-D5
CAS 2909407-29-8
Molekylær formel C15 D5 H5 O6
Mængde 0.01 g
Synonym 2-(3,4-Dihydroxyphenyl-2,5,6-d3)-3,7-dihydroxy-4 H-chromen-4-one-6,8-d2, Fisetin-d5, Fisetin-2',5',6,6',8-d5, Fisetin-2',5',6,6',8-D5
InChI formel InChI=1 S/C15H10O6/c16-8-2-3-9-12(6-8)21-15(14(20)13(9)19)7-1-4-10(17)11(18)5-7/h1-6,16-18,20 H/i1D,2 D,4 D,5 D,6 D
SMIL [2 H]c1cc2C(=O)C(=C(Oc2c([2 H])c1O)c3c([2 H])c([2 H])c(O)c(O)c3[2 H])O
IUPAC navn 6,8-dideuterio-3,7-dihydroxy-2-(2,3,6-trideuterio-4,5-dihydroxyphenyl)chromen-4-one
Molekylvægt (g/mol) 291.2671
Formel vægt 291.0791 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity