Få mere at vide
Firocoxib-d4, TRC
Artikelnummer. 30403560
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30403560 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403560 Leverandør Toronto Research Chemicals Leverandørnr. TRCF3560021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Firocoxib cyclopropyl-2,2,3,3-d4
CAS 1325700-11-5
Renhedsgrad noter HPLC
Molekylær formel C17 2H4 H16 O5 S
Mængde 1 mg
Synonym 5,5-Dimethyl-4-(4-methylsulfonylphenyl)-3-[(cyclopropyl-2,2,3,3-d4)methoxy]furan-2-one, 5,5-Dimethyl-4-(4-methylsulfonylphenyl)-3-[(2,2,3,3-tetradeuteriocyclopropyl)methoxy]furan-2-one, Firocoxib-d4, Firocoxib-D4 (cyclopropyl-2,2,3,3-D4)
InChI formel InChI=1S/C17H20O5S/c1-17(2)14(12-6-8-13(9-7-12)23(3,19)20)15(16(18)22-17)21-10-11-4-5-11/h6-9,11H,4-5,10H2,1-3H3/i4D2,5D2
SMIL [2H]C1([2H])C(COC2=C(c3ccc(cc3)S(=O)(=O)C)C(C)(C)OC2=O)C1([2H])[2H]
IUPAC navn 5,5-dimethyl-4-(4-methylsulfonylphenyl)-3-[(2,2,3,3-tetradeuteriocyclopropyl)methoxy]furan-2-one
Molekylvægt (g/mol) 340.43
Formel vægt 340.1283
Procent renhed >95