Få mere at vide
Fingolimod-d4 Hydrochloride, TRC
Artikelnummer. 30406041
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30406041 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406041 Leverandør Toronto Research Chemicals Leverandørnr. TRCF80500225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fingolimod-d4 Hydrochloride
CAS 1346604-90-7
Molekylær formel C19H30D4ClNO2
Mængde 25 mg
Synonym 2-Amino-2-(2-(4-octylphenyl)ethyl)propane-1,3-diol-d4 Hydrochloride, FTY720-d4 Hydrochloride
InChI formel InChI=1S/C19H33NO2.ClH/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-22;/h9-12,21-22H,2-8,13-16,20H2,1H3;1H/i15D2,16D2;
SMIL C(CC(C(O)([2H])[2H])(C(O)([2H])[2H])N)C1=CC=C(CCCCCCCC)C=C1.Cl
IUPAC navn 2-amino-2-(4-octylphenethyl)propane-1,1,3,3-d4-1,3-diol hydrochloride
Molekylvægt (g/mol) 347.96
Formel vægt 347.25