Få mere at vide
Fingolimod-d4 HCl (1,3-propane-1,1,3,3-d4-diol), CDN
Artikelnummer. 30317496
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317496 0.005 g
30316216 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317496 Leverandør CDN Leverandørnr. CDND79710.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fingolimod-d4 Hydrochloride
CAS 1346604-90-7
Molekylær formel C19H30D4ClNO2
Mængde 0.005 g
Synonym 2-Amino-2-(2-(4-octylphenyl)ethyl)propane-1,3-diol-d4 Hydrochloride, FTY720-d4 Hydrochloride;
InChI formel InChI=1 S/C19H33NO2.ClH/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-22;/h9-12,21-22 H,2-8,13-16,20H2,1H3;1 H/i15D2,16D2;
SMIL C(CC(C(O)([2 H])[2 H])(C(O)([2 H])[2 H])N)C1=CC=C(CCCCCCCC)C=C1.Cl
IUPAC navn 2-amino-2-(4-octylphenethyl)propane-1,1,3,3-d4-1,3-diol hydrochloride
Molekylvægt (g/mol) 347.96
Formel vægt 347.25 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity