Learn More
KNF Neuberger™ Aluminum/PTFE Vacuum Gauge Valve
Brand:  KNF Neuberger™ 000350
Additional Details : Weight : 0.50000kg
Product Code. 10314781
  • 630.00 DKK / Each
Estimated Shipment: 11-06-2024
to see stock.
Specifications
Aluminum, PTFE
20.3 cm
Vacuum Gauge Valve
194 mm
1 Each
4 kg