Learn More
Thermo Scientific™ Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Multiskan™ FC microplate photometers can be equipped with wide range of additional filters to expand the instrument functionality and application flexibility.
1936.00 DKK - 7150.00 DKK
Specifications
Product Type Optical Filter
For Use With (Equipment) Multiskan FC
View More Specs
Products 20
Product Code Brand Product Type Wavelength Price Quantity & Availability  
Product Code Brand Product Type Wavelength Price Quantity & Availability  
11889530
View Documents Promotion Details
Thermo Scientific™
1423405
Optical Filter 340 nm
1942.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
11540705
View Documents Promotion Details
Thermo Scientific™
1423755
Optical Filter 375 nm
5610.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
11709458
View Documents Promotion Details
Thermo Scientific™
1424055
Optical Filter 405 nm
1936.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
11719458
View Documents Promotion Details
Thermo Scientific™
1424145
Optical Filter 414 nm
1938.00 DKK
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11729458
View Documents Promotion Details
Thermo Scientific™
1424505
Optical Filter 450 nm
1940.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
11799448
View Documents Promotion Details
Thermo Scientific™
1424925
Optical Filter 492 nm
1938.00 DKK
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
12566528
View Documents Promotion Details
Thermo Scientific™
1425205
Optical Filter 520 nm
5450.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
11560705
View Documents Promotion Details
Thermo Scientific™
1425305
Optical Filter 530 nm
7020.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
11755058
View Documents Promotion Details
Thermo Scientific™
1425405
Optical Filter 540 nm
1940.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
11709508
View Documents Promotion Details
Thermo Scientific™
1425505
Optical Filter 550 nm
1940.00 DKK
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11761218
View Documents Promotion Details
Thermo Scientific™
1425605
Optical Filter 560 nm
5550.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
11794968
View Documents Promotion Details
Thermo Scientific™
1425705
Optical Filter 570 nm
1938.00 DKK
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11829540
View Documents Promotion Details
Thermo Scientific™
1425955
Optical Filter 595 nm
1940.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 03-07-2024
to see stock.
11500705
View Documents Promotion Details
Thermo Scientific™
1426205
Optical Filter 620 nm
1966.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
11530705
View Documents Promotion Details
Thermo Scientific™
1426305
Optical Filter 630 nm
3865.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-06-2024
to see stock.
11765058
View Documents Promotion Details
Thermo Scientific™
1426505
Optical Filter 650 nm
1938.00 DKK
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11782278
View Documents Promotion Details
Thermo Scientific™
1426905
Optical Filter 690 nm
1940.00 DKK
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11580705
View Documents Promotion Details
Thermo Scientific™
1427405
Optical Filter 740 nm
5590.00 DKK
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11713869
View Documents Promotion Details
Thermo Scientific™
1427505
Optical Filter 750 nm
4660.00 DKK
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11500715
View Documents Promotion Details
Thermo Scientific™
1428205
Optical Filter 820 nm
7150.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 03-07-2024
to see stock.
Description
Description
Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.
  • Multiskan FC is equipped with an eight-position filter wheel with three preinstalled standard filters, 405 nm, 450 nm and 620 m.
  • A comprehensive range of easy-to install additional filters is available for order
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
Specifications
Specifications
Optical Filter
Multiskan FC