Learn More
Hamilton™ Filtered Pre-Sterilized Racked Pipette Tips
Delivering outstanding accuracy and precision, guaranteed.
1390.00 DKK - 1490.00 DKK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15345084
|
Hamilton™
68009-13 |
300 μL |
1490.00 DKK
Pack of 10 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
15375094
|
Hamilton™
68007-13 |
10 μL |
1390.00 DKK
Pack of 10 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
Description
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Pre-sterilized
- 96 tips to a rack
- Standard retention
Specifications
Filtered | |
Sterile | |
Polypropylene |