Learn More
Hamilton™ Filtered Low Retention Pre-Sterilized Racked Pipette Tips
With surface walls completely free of occlusions and cavities so fluid retention is virtually eliminated.
1320.00 DKK - 1580.00 DKK
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Format | Rack |
Material | Polypropylene |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15315084
|
Hamilton™
68009-23 |
300 μL |
1575.00 DKK
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
15335094
|
Hamilton™
68007-23 |
10 μL |
1580.00 DKK
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
15375084
|
Hamilton™
68008-23 |
200 μL |
1320.00 DKK
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Pre-sterilized
- 96 tips to a rack
- Delivering outstanding accuracy and precision, guaranteed
Specifications
Filtered | |
Sterile | |
Rack | |
Polypropylene |