missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Filter Funnels

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 07195479

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10111431

  • 6370.00 DKK / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

  • Filter funnels
  • Material: borosilicate glass 3.3
Specifications

Specifications

4 L
400 mm
4
Borosilicate Glass 3.3
175 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers