missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 50 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 15501

 View more versions of this product

Product Code. 12050432

  • 590.00 DKK / Pack of 10
Estimated Shipment: 04-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
Non-sterile
1
50 mm
1 μm
10/Pk.
4.5 ± 1.0 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers