missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 170 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 51700

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12023998

  • 1251.00 DKK / Each


Estimated Shipment
27-05-2021
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

170 mm
13.0 ± 3.0 mm
Non-sterile
1 Ea.
Borosilicate Glass 3.3
Videos
SDS
Documents