missing translation for 'onlineSavingsMsg'
Learn More

Filter aid, Harborlite 800

CAS: 130885-09-5

348.00 DKK - 1060.00 DKK

Chemical Identifiers

CAS 130885-09-5
MDL Number MFCD00197888
Synonym Perlite, filter aid; Harborlite J2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11303177
SDS View Documents
Thermo Scientific Alfa Aesar
L13608.36
500 g
348.00 DKK
500g
Please to purchase this item. Need a web account? Register with us today!
11313177
SDS View Documents
Thermo Scientific Alfa Aesar
L13608.0E
2500 g
1060.00 DKK
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

Solubility
Insoluble in water.

Notes
Harborlite is a registered trademark of Harborlite Corportion.
Specifications

Chemical Identifiers

130885-09-5
MFCD00197888
Perlite, filter aid; Harborlite J2

Specifications

2.3
MFCD00197888
Insoluble in water.
130885-09-5
Perlite, filter aid; Harborlite J2
Filter aid, Harborlite 800
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'rtecsNumber' : VV7311000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.