missing translation for 'onlineSavingsMsg'
Learn More

Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined

CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol

476.00 DKK - 1210.00 DKK

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15498335
SDS View Documents
Thermo Scientific Alfa Aesar
H33152.0B
1000 g
476.00 DKK
1000g
Please to purchase this item. Need a web account? Register with us today!
15408345
SDS View Documents
Thermo Scientific Alfa Aesar
H33152.0I
5000 g
1210.00 DKK
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong acids.
Specifications

Chemical Identifiers

68855-54-9
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
CHEBI:29377
C(=O)([O-])[O-].[Na+].[Na+]
CNa2O3
MFCD00132803
10340
disodium;carbonate

Specifications

68855-54-9
MFCD00132803
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
Filter aid, Celite™ 545, treated with sodium carbonate, flux calcined
CNa2O3
Insoluble in water.
C(=O)([O-])[O-].[Na+].[Na+]
105.988
CHEBI:29377
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H373-H319-H335
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 272-489-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.