Learn More
Cytiva Filter
Brand:  Cytiva 10409872
1401.25 DKK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 11999217
  • 1475.00 DKK / Pack of 100
Estimated Shipment: 20-08-2024
to see stock.
Specifications
5 cm dia.
5 cm
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose