Få mere at vide
Tocris Bioscience™ Filipin III
Polyene antibiotic; inhibits formation of pathological isoform of prion protein
Brand: Tocris Bioscience™ 6250/1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Filipin III is a polyene antibiotic. Binds sterols in fungal membranes. Also inhibits formation of the pathological form of prion protein (PrP-res) from the normal membrane-bound (PrP-sen) form. Also fluorescent cholesterol stain.Tekniske data
| Polyene Antibiotic, Inhibits Formation of Pathological Isoform of Prion Protein | |
| C35H58O11 | |
| Store at -20°C | |
| 1 mg |
| 480-49-9 | |
| 654.83g/mol | |
| Antibiotics |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only