missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Activation Protein alpha/FAP Antibody (FAP/4851), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14219JF646
Additional Details : Weight : 0.00970kg
Description
Fibroblast Activation Protein alpha/FAP Monoclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Fibroblast Activation Protein alpha/FAP | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2c κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
FAP/4851 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
Recombinant fragment (around aa 1-200) of human Fibroblast Activation Protein alpha/FAP protein (exact sequence is proprietary) (Uniprot: Q12884) | |
0.1 mL | |
Cancer, Protein Phosphatase | |
2191 | |
Store at 4°C in the dark. | |
Purified |