Learn More
Thermo Scientific Chemicals Fibrinopeptide B, Human
For cellular and molecular biology applications
Brand:  Thermo Scientific Chemicals J66812.#0
Additional Details : CAS Number : 36204-23-6 Weight : 0.00005kg
 View more versions of this product
Product Code. 15478319
  • 3293.81 DKK / 5mg
Estimated Shipment: 24-05-2024
to see stock.
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Human
-30°C to -10°C
1552.55
5 mg
Glp-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg
HPLC
Solid
Fibrinopeptide B, human
RUO - research use only
Product Suggestions
Product Suggestions
Videos
SDS Documents