missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FHL5 Recombinant Protein

Brand:  Abnova™ H00009457-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16143692

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH21723
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
56.98
8
Glutathione Sepharose 4 Fast Flow
25 ug
MTTAHFYCQYCTASLLGKKYVLKDDSPYCVTCYDRVFSNYCEECKKPIESDSKDLCYKDRHWHEGCFKCTKCNHSLVEKPFAAKDERLLCTECYSNECSSKCFHCKRTIMPGSRKMEFKGNYWHETCFVCENCRQPIGTKPLISKESGNYCVPCFEKEFAHYCNFCKKVITSGGITFCDQLWHKECFLCSDCRKDLCEEQFMSRDDYPFCMDCYNHLYANKCVACSKPISGLTGAKFICFQDSQWHSECFNCGKCSVSLVGKGFLTQNKEIFCQKCGSGMDTDI
ACT/FLJ33049/KIAA0776/RP3-393D12.2/dJ393D12.2
FHL5
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
9457
FHL5 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FHL5
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers